Would you like to log in again?
Sepharose™ CL-4B Phenyl Hydrophobic Interaction Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sepharose™
17-0810-01 17-0810-01
95016-996EA 838.85 USD InStock
CA95016-996L 95016-996
Sepharose™ CL-4B Phenyl Hydrophobic Interaction Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Octyl-Sepharose™ CL-4B is a well-proven medium for hydrophobic interaction chromatography (HIC). Octyl immobilised on 4% cross-linked agarose.
  • Phenyl immobilized on 4% cross-linked agarose
  • Aromatic ligand
  • Stable in all commonly used aqueous buffers and additives such as 8 M urea and 6 M guanidine hydrochloride
  • Negligible volume variation due to changes in pH or ionic strength
Octyl Sepharose® CL-4B and Phenyl Sepharose® CL-4B are covalently coupled to a cross-linked 4% agarose matrix by ether linkage, resulting in hydrophobic gels with minimal leakage and no ionic properties.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty