Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

illustra NAP Columns, Cytiva

Supplier: Cytiva

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

illustra™
17-0852-02 17-0853-01 17-0852-02 17-0853-02 17-0853-02 17-0853-01 17-0854-02 17-0854-01 17-0854-01 17-0852-01 17-0854-02 17-0852-01
95017-013PK 648.23 USD InStock
95017-013 CA95017-005L 95017-011 CA95017-013L CA95017-003L CA95017-011L 95017-009 95017-007 95017-005 CA95017-009L CA95017-007L 95017-003
illustra NAP Columns, Cytiva
Chromatography Columns
NAP Columns are disposable columns, prepacked with Sephadex G-25 DNA Grade, that require only gravity to run.

  • For rapid and efficient purification of DNA and oligonucleotides (10-mers) in less than 15 min by gravity flow
  • Prepacked with Sephadex G-25 DNA Grade in distilled water containing 0.15% Kathon CG/ICP Biocide
  • Available in three sizes depending on sample volume: 0.5 ml (NAP-5), 1 ml (NAP-10) or 2.5 ml (NAP-25)
  • Useful for small-scale purification, desalting and buffer exchange

Delivery information: Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration, washing and sample elution.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty