Would you like to log in again?
Affinity purification media, GammaBind™ Plus Sepharose™
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sepharose™
17-0886-02 17-0886-01 17-0886-01 17-0886-02 17-0886-04 17-0886-04
95017-024EA 3555.74 USD InStock
95017-024 95056-150 CA95017-024L 95017-022 CA95017-022L CA95056-150L
Affinity purification media, GammaBind™ Plus Sepharose™
Protein Purification Protein Purification Resins
GammaBind Plus Sepharose is GammaBind G, Type 3, covalently immobilized to Sepharose CL-6B by malimide linkage.
  • For affinity chromatography
  • Recombinant Protein G type 3 coupled to Sepharose CL-6B for enhanced binding to rat and mouse IgG subclasses at laboratory scale
  • Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
  • Maleimide activation coupling method
  • Yields up to 18 mg/mL mouse IgG
  • Stable in all commonly used aqueous buffers
GammaBind G, Type 3, a recombinant form of streptococcal protein G, binds to the Fc region of IgG from a variety of mammalian species. GammaBind Plus Sepharose may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium. Since only the Fc region is involved in binding, the Fab region is still available for binding antigen.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty