Would you like to log in again?
IMAC Sepharose™ 6 Fast Flow Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17092107 17-0921-07 17-0921-08 17-0921-08 17-0921-08
97067-852EA 365.42 USD InStock
CA95017-034 97067-852 95017-034 CA97067-852 97067-854
IMAC Sepharose™ 6 Fast Flow Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
IMAC Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix.
  • For affinity chromatography
  • Uncharged IMAC resin allowing possibility to charge with your metal ion of choice for optimized selectivity
  • Convenient purification of histidine-tagged proteins, also when Ni2+ is not the best choice of metal ion
  • Stable in commonly used aqueous buffers 0.01 M HCl, 1 M NaOH, 70% acetic acid, 2% SDS. 30% 2-propanol
IMAC Sepharose 6 Fast Flow is a member of the Cytiva range of IMAC chromatography media for capture and intermediate purification. As a BioProcess medium, IMAC Sepharose 6 Fast Flow fulfills industrial demands for security of supply, robust performance and regulatory support. The coupling chemistry and the novel ligand is designed to give minimal ligand leakage while providing high capacity.
IMAC Sepharose 6 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, IMAC Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
IMAC Sepharose 6 Fast Flow is supplied free of metal ions, allowing the user to charge it with the most appropriate metal ion for purification of a target protein
Ordering information: Available in the convenient and time-saving prepacked HiTrap and HiPrep formats.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty