Would you like to log in again?
Protein A Sepharose™ CL-4B Affinity Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sepharose™
17-0963-02 17-0780-01 17-0963-03 17-0780-01 17-0963-03 17-0963-02
95017-044EA 2534.53 USD InStock
95017-044 CA95055-962L CA95017-044L CA95016-992L 95055-962 95016-992
Protein A Sepharose™ CL-4B Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Protein A binds to the Fc region of immunoglobulins through interactions with the heavy chain.
  • For affinity chromatography
  • For immunoglobin purification and fractionation, immunoprecipitation procedures and polyclonal antibody purification
  • Strong binding to Fc region of IgG
  • High capacity - 20 mg human IgG/ml
  • Cyanogen bromide activation coupling method
  • Stable in all commonly used aqueous buffers, 6 M guanidine-HCl, 8 M urea
  • Freeze-dried powder or preswollen in 20% ethanol
  • Negligible volume variation due to changes in pH or ionic strength
The binding of protein A has been well documented for IgG from a variety of mammalian species and for some IgM and IgA as well. Protein A Sepharose CL-4B has been used as a powerful tool to isolate and purify classes, subclasses and fragments of immunoglobulins from biological fluids and from cell culture media. Since only the Fc region is involved in binding, the Fab region is available for binding antigen.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty