Would you like to log in again?
IgG Sepharose™ 6 Fast Flow Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17-0969-01 17-0969-01
95017-050EA 629.27 USD InStock
95017-050 CA95017-050L
IgG Sepharose™ 6 Fast Flow Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
lgG Sepharose™ 6 Fast Flow, from Cytiva, is based on the rigid Sepharose 6 Fast Flow matrix, with human IgG covalently coupled to it.
  • For affinity chromatography
  • Recombinant tagged proteins containing a protein A tag
  • Binds at least 2 mg protein A/ml medium
  • Suitable for tandem affinity purification (TAP) of protein complexes
  • Stable to all commonly used aqueous buffers
The improved mechanical characteristics of this Fast Flow medium allows high flow rates to be used for rapid and convenient single step purification of protein A fusion protein conjungates produced in prokaryotic expression systems. IgG coupled to the highly cross-linked 6% agarose matrix Sepharose 6 Fast Flow, designed for the rapid purification of protein A and protein A fusion conjugates.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty