Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

Supplier: Cytiva
Warning
Synonyms: Trizma(R) baseTRIS baseTHAM2-Amino-2-(hydroxymethyl)-1,3-propanediolTris(hydroxymethyl)methylamine2-Amino-2-(hydroxymethyl)propane-1,3-diolTromethamine

17-1321-01 17-1321-01
95017-134EA 144.63 USD InStock
95017-134 CA95017-134L
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Tris (tris[hydroxymethyl] aminoethane) is suitable for preparing electrophoresis buffer in the pH range of 7,2 to 9,0. It is qualified for 2-D electrophoresis.


Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Melting Pt: 169…172 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 12,09902
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/00/2297600.jpg
Order Now
Specification Test Results
Application For preparing electrophoresis buffer. Used in PAGE and Sequencing.
Purity ≥99,8%
pH Range 7.2 - 9.0
Boiling/Condensation Point 219 - 220°C
Melting/Freezing Point 170°C
Solubility Soluble in cold water and hot water, methanol
Physical State Solid
Color White
Storage conditions 10 - 30°C
Estimated shelf life from manufacture date 12 month
Wetted Part No
Guidance Tris = (tris(hydroximethyl)aminoethane)
DIGE approved Y

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty