Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PlusOne™ Repel-Silane ES, colorless liquid

Supplier: Cytiva
Warning

17-1332-01 17-1332-01
95017-150EA 193.88 USD InStock
95017-150 CA95017-150L
PlusOne™ Repel-Silane ES, colorless liquid
PlusOne™ Repel-Silane ES
Repel-Silane ES is a 2% solution of dimethyldichlorosilane dissolved in octamethylcyclooctasilane.

  • Used in IEF, PAGE and Sequencing
  • Soluble in cold water and hot water

It is used to inhibit sticking of polyacrylamide gels, agarose gels, and nucleic acids to glass surfaces.


Flash Pt: 56 °C (132.8 °F) (closed cup)
Storage Temperature: Ambient
UN: 1993
ADR: 3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty