Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Precast ExcelGel for SDS-PAGE, Cytiva
Supplier: Cytiva
ExcelGel®
17-1342-01 80-1261-01 17-1342-01 80-1261-01
95017-153PK 212.86 USD InStock
95017-153 CA95017-153L CA95017-751L 95017-751
Precast ExcelGel for SDS-PAGE, Cytiva
Electrophoresis Gels Precast Gels
ExcelGel precast gels for SDS-PAGE are available as homogeneous or gradient gels. The homogeneous gels have 25 preformed sample wells for sample volumes from 5 to 17 µl. The gradient gels use sample application strips (26 samples up to 40 µl, 52 samples up to 20 µl), paper pieces or sample droplets. All ExcelGel SDS homogeneous and 8 - 18 gradient gels have a 33 mm stacking zone and a 77 mm separating zone.
  • Save time with ready-to-use, mechanically stable gels cast on plastic supports
  • Apply samples quickly and easily using preformed sample wells
  • Avoid making running buffer by using precast buffer strips
ExcelGel gradient gels are recommended for use with Immobiline DryStrip gels.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty