Would you like to log in again?
Streptavidin Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sepharose™
17-5113-01 17-5113-01
95017-166EA 709.71 USD InStock
95017-166 CA95017-166L
Streptavidin Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Streptavidin Sepharose™ High Performance is an affinity chromatography medium designed for fast and reliable, high resolution purification of biotinylated biomolecules. Purified Streptavidin isolated from Streptomyces avidinii is immobilised on Sepharose™ High Performance.
  • Extremely useful for exploiting either the strong interactions of biotin and streptavidin or the somewhat weaker interactions of 2-iminobiotin and streptavidin
  • 34 µm bead size for improved resolution
The base matrix is a rigid, highly crosslinked beaded agarose with high chemical stability. The immobilised streptavidin binds biotin and biotinylated substances through affinity chromatography.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty