Would you like to log in again?
Benzamidine Sepharose™ 4 Fast Flow (High Sub) Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17-5123-10 17-5123-10
95017-170EA 536.37 USD InStock
95017-170 CA95017-170L
Benzamidine Sepharose™ 4 Fast Flow (High Sub) Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Benzamidine Sepharose 4 Fast Flow (high sub) is an excellent tool for removal and/or purification of trypsin and trypsin-like serine proteases, as well as, zymogens including urokinase and prekallikrein
  • For affinity chromatography
  • p-aminobenzamidine (pABA) immobilized on Sepharose 4 Fast Flow matrix gives high productivity, and allows easy scale-up
  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
  • Amide coupling method
  • Stable in all commonly used aqueous buffers
Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates. Benzamidine Sepharose 4 Fast Flow (high sub) is based on highly cross-linked 4% agarose enabling high flow rates without losing binding capacity
For convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases, and zymogens including urokinase and prekallikrein.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty