Would you like to log in again?
Glutathione Sepharose™ 4 Fast Flow Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17-5132-02 17-5132-02 17-5132-03 17-5132-01 17-5132-03 17-5132-01
95017-176EA 9178.38 USD InStock
95017-176 95017-174 CA95017-174L 95017-172 CA95017-176L CA95017-172L
Glutathione Sepharose™ 4 Fast Flow Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Glutathione Sepharose 4 Fast Flow is used for the purification of GST-tagged proteins from bacterial lysates and other S-tranferases or glutathione-dependent proteins.
  • For affinity chromatography
  • Fast, one-step purification of GST-tagged proteins
  • Mild elution conditions preserving protein antigenicity and function
  • For scale-up due to high flow properties
  • Glutathione coupled to Sepharose 4 Fast Flow via a 10-carbon spacer arm
  • Epoxy activation coupling method
  • Stable in all commonly used aqueous buffers
Mild elution conditions preserve protein antigenicity and function. Sepharose 4 Fast Flow provides good flow properties and is excellent for scaling up. Binding of GST to glutathione is flow-rate dependent and lower flow rates often increase the binding capacity. This is important to consider during sample loading and elution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty