Would you like to log in again?
Ni Sepharose™ 6 Fast Flow Affinity Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sepharose™
17-5318-02 17-5318-01 17-5318-03 17-5318-06 17531802 17-5318-03 17-5318-01 17-5318-01 17-5318-06 17-5318-06
95017-178EA 450.9 USD InStock
95017-178 CA95017-178 97067-850 97067-844 95017-184 95017-182 CA97067-846 97067-846 CA95017-182 CA95017-184
Ni Sepharose™ 6 Fast Flow Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Ni Sepharose 6 Fast Flow is a BioProcess medium that combines the advantages of Ni₂₊ for purification of histidine tagged proteins, with the well industrially established properties of the Sepharose Fast Flow platform.
  • Designed for scaling up purification of histidine-tagged proteins
  • High protein binding capacity and minimal leakage of Ni2₊ ions
  • ~15 µmol Ni₂₊/mL medium metal ion capacity
  • As BioProcess medium it fulfills industrial demands (security, performance and regulatory)
  • Available in convenient prepacked HisPrep and HisTrap column formats
  • Stable in commonly used aqueous buffers - 0.1 M HCl, 0.1 M NaOH, 8 M urea
The coupling chemistry and the novel ligand is designed to give minimal ligand and Ni2₊ leakage while providing high capacity. Ni Sepharose 6 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Ni Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Ni Sepharose 6 Fast Flow is a member of the Cytiva range of IMAC chromatography media for capture and intermediate purification. Ni Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix and pre-charged with Ni2₊ ions for purification of histidine tagged proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty