Would you like to log in again?
Streptavidin, Cy3®, Cytiva
Supplier: Cytiva
CY™
PA43001 PA43001
95017-402EA 340.01 USD InStock
95017-402 CA95017-402L
Streptavidin, Cy3®, Cytiva
Proteins and Peptides
Streptavidin conjugated to a range of fluorescent dyes for use in in situ hybridization and flow cytometry.
  • High specificity reagents
This product is supplied as a lyophilized solid in phosphate-buffered saline (0.02 M potassium phosphate, 0.15 M NaCl) pH 7.2, containing 1%(w/v) bovine serum albumin and 0.1% (w/v) sodium azide. Reconstitute with 1.0 ml of deionized water, dispense into suitable aliquots and freeze. Avoid repeated freeze-thaw cycles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty