Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nucleon BACC Genomic DNA Extraction Kits, Cytiva

Supplier: Cytiva
RPN8501 RPN8501 RPN8512 RPN8512
95017-486EA 544.88 USD InStock
95017-486 CA95017-486L 95017-484 CA95017-484L
Nucleon BACC Genomic DNA Extraction Kits, Cytiva
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Nucleon™ BACC Genomic DNA Extraction Kits were developed for rapid extraction of high quality, high molecular weight genomic DNA from blood and cell cultures.

  • Developed for rapid extraction of high-quality, high molecular weight genomic DNA from whole blood and cell cultures
  • Recovers DNA ranging in size from 23 to 250 kbp as determined by pulse-field gel electrophoresis
  • Features a phenol-free protocol requiring only 30 min to complete

The Nucleon™ proprietary resin is added following cell lysis, deproteinisation with sodium perchlorate, and a single chloroform extraction. Aspiration of the top layer of nucleic acids is simplified and volume of usable sample maximized by a barrier formed between the two layers.

Parameter
Application: Rapid extraction of high-quality, high molecular weight genomic DNA from cell cultures and blood.
Protocol length: 30 min
Sample type: Cell cultures and blood
Yield: 370-440 µg DNA/mg tissue
Mean Purity (A 260/280): 1.8

Nucleon™ BACC1: For 50 preps of 1 ml of whole blood or cultured cells (1 to 3×10⁶ cells/ml)

Nucleon™ BACC3: For 50 preps of 10 ml of whole blood (kit contains all reagents necessary for 50×10 ml blood extractions).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty