Would you like to log in again?
Amersham™ CyDye™ Mono-Reactive NHS Esters, Cytiva
Supplier: Cytiva
CY™
PA15604 PA15606 PA17101 PA13106 PA15104 PA15601 PA15106 PA15102 PA15604 PA15606 PA15100 PA15602 PA13104 PA15100 PA13105 PA13101 PA15605 PA17101 PA15106 PA15101 PA15101 PA17104 PA15601 PA13104 PA15602 PA17105 PA13105 PA15104 PA17104 PA15605 PA15102 PA13101
95017-354EA 3128.43 USD InStock
CA95017-354L CA95017-540L 95017-354 95017-352 CA95017-544L CA95017-350L 95017-350 CA95017-362L 95056-514 CA95017-352L CA95017-342L CA95017-548L CA95017-524L CA95017-502L CA95017-508L CA95017-528L CA95017-506L CA95017-538L 95017-508 95017-528 95017-506 95017-526 95017-548 CA95056-514L 95017-546 95017-524 95017-502 95017-544 95017-342 95017-540 95017-362 95017-538
Amersham™ CyDye™ Mono-Reactive NHS Esters, Cytiva
Protein Linking and Labeling Systems
Mono-reactive dye pack contains derivatised CyDye™ that carries only one reactive group on each dye molecule for targeted and specific labeling of amine groups, for example on antibodies, peptides or oligonucleotides.
Delivery information: Example protocols included with each dye pack.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty