Would you like to log in again?
Amersham Cy3 and Cy5 Maleimide Mono-Reactive Dye 5-Packs, Cytiva
Supplier: Cytiva
CY™
PA25031 PA23031 PA23031 PA25031
95017-551PK 1118.45 USD InStock
CA95017-551L 95017-551 CA95017-553L 95017-553
Amersham Cy3 and Cy5 Maleimide Mono-Reactive Dye 5-Packs, Cytiva
Protein Linking and Labeling Systems
Mono-functional maleimides are particularly suitable for the selective labelling of molecules containing free sulfhydryl groups, such as cysteine residues in proteins and peptides and oligonucleotides.
  • More selective method of antibody labelling that reduces the possibility of the label interfering with subsequent antibody-antigen reactions
  • Provides an alternative when CyDye™ NHS esters are unsuitable
  • Dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labelings
They are also useful in antibody labelling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.
Delivery information: Each kit contains five vials of dye, each sufficient for labelling 1 mg of protein.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty