Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Cytiva
NXA931-1ML NXA931-1ML
95017-554EA 454.8 USD InStock
CA95017-554L 95017-554
Anti-IgG Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Highly species specific HRP-conjugated antibodies optimized for use with Amersham ECL Western Blotting Detection Reagents.

Optimized for use with our range of Amersham ECL Western blotting detection reagents.

The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2°C to 8°C.

Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: none
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty