Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PreScission™ Protease Affinity Chromatography Media, Cytiva

Supplier: Cytiva
Prescission™
27-0843-01 27-0843-01
95017-584EA 591.1 USD InStock
CA95017-584L 95017-584
PreScission™ Protease Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Enables the low-temperature cleavage of fusion proteins containing the PreScission Protease recognition sequence.

  • Cleavage buffer 50 mM Tris-HCl, 150 mM NaCl, 1 mM EDTA, 1 mM dithiothreitol (DTT), pH 7.0
  • Elution buffer 50 mM Tris-HCl, 10 mM reduced glutathione, pH 8.0
  • Inhibitor 100 mM ZnCl2 (\< 50% inhibition); 100 µM chymostatin; 4 mM Pefabloc

Can be used either following affinity purification or while fusion proteins are bound to Glutathione Sepharose 4B Glutathione Sepharose High Performance, or Glutathione Sepharose 4 Fast Flow.

PreScission Protease is a genetically engineered fusion protein consisting of human rhinovirus 3C protease and GST. This protease was designed to facilitate removal of the protease by allowing simultaneous protease immobilization and cleavage of GST fusion proteins produced from the pGEX-6P vectors pGEX-6P-1, pGEX-6P-2, and pGEX-6P-3; see pGEX Vectors (GST Gene Fusion System). PreScission Protease specifically cleaves between the Gln and Gly residues of the recognition sequence of LeuGluValLeuPheGln/GlyPro.

One unit will cleave > 90% of 100 µg of a test GST-fusion protein at 5°C for 16 hours.

Ordering information: 2000 units/ml (833 to 1000 units/mg) in 50 mM Tris-HCl (pH 8.0), 150 mM NaCl, 10 mM EDTA, 1 mM DTT, and 20% glycerol.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty