Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thrombin

Supplier: Cytiva
27-0846-01 27-0846-01
95017-586EA 220.63 USD InStock
CA95017-586L 95017-586
Thrombin
Enzymes
Thrombin is a protease used to digest fusion proteins prepared from pGEX vectors containing the recognition sequence for thrombin (pGEX-1lT, pGEX-2T, pGEX-2TK, pGEX-4T-1, pGEX-4T-2, and pGEX-4T-3).

  • Enables the site-specific cleavage of fusion proteins containing an accessible thrombin recognition sequence
  • Purified from bovine plasma; functionally free of other clotting factors, plasminogen, and plasmin
  • Can be used either following affinity purification or while fusion proteins are bound to Glutathione Sepharose™ 4B, Glutathione Sepharose™ High Performance, or Glutathione Sepharose™ 4 Fast Flow

Application: Used for site-specific separation of the GST tag from proteins expressed using pGEX-T vectors.

Cleavage Buffer: PBS 140 mM NaCl, 2,7 mM KCl, 10 mM Na2HPO4, 1,8 mM KH2PO4, pH 7.3
Elution Buffer:50 mM Tris-HCl, 10 mM reduced glutathione, pH 8.0

Inhibitors: Pefabloc TH benzamidine, AEBSF, APMSF, antithrombin III, Antipain, α1-antitrypsin, aprotinin, chymostatin, hirudin, leupeptin, PMSF

Molecular Weight: 37000

Unit Definition: One unit of enzyme cleaves 90% of 100 μg of a test GST fusion protein when incubated in 1X PBS at 22 °C for 16 hours

Ordering information: Supplied as lyophilized powder. Includes 50 mM Tris-HCl, 10 mM reduced glutathione, pH 8.0.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty