Would you like to log in again?
Disposable Columns Prepacked with Sepharose™, GST Bulk Kit, Cytiva
Supplier: Cytiva
27-4570-01 27-4570-01
95017-598EA 1214.85 USD InStock
CA95017-598L 95017-598
Disposable Columns Prepacked with Sepharose™, GST Bulk Kit, Cytiva
Chromatography Columns
GST bulk kit is designed for small-scale manual purification of GST-tagged proteins using either column chromatography or batch method. Facilitates optimization studies of expression, solubility conditions and purification parameters of GST-tagged proteins.
  • For rapid purification of GST-tagged proteins using gravity flow columns or using batch purification with high specificity
  • Preserves protein antigenicity and function
  • Yields up to 50 mg of pure protein
GST-tagged proteins can be recovered under mild elution conditions (10 mM glutathione), which preserves functionality of the proteins. The binding capacity of Glutathione Sepharose 4B, which can be greater than 10 mg protein/ml medium, varies with the nature of the tagged protein and the exact binding conditions used.
GST Bulk Kit is designed for small-scale manual purification of GST-tagged proteins using either column chromatography or a batch method. The batch method is more flexible, as purification can be performed with between 10 μl and 2 ml of Glutathione Sepharose 4B.
GST Bulk Kit also facilitates optimization studies of expression, solubility conditions, and purification parameters of GST-tagged proteins.
Ordering information: Includes 10 ml bulk pack of Glutathione Sepharose 4B medium, 5× gravity-flow columns and IPTG GST buffer kit. The kit contains medium and reagents for purification of GST-tagged proteins from milliliter volumes of sample to larger volumes of sample containing as much as 50 mg of GST-tagged protein.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty