Would you like to log in again?
illustra™ MicroSpin™ G-50 columns, Cytiva
Supplier: Cytiva
illustra™ MicroSpin™
27-5330-02 27-5330-02 27-5120-01 27-5330-01 27-5120-01 27-5330-01
95017-615PK 409.4 USD InStock
CA95017-625L 95017-615 95017-625 CA95017-623L 95017-623 CA95017-615L
illustra™ MicroSpin™ G-50 columns, Cytiva
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
For rapid purification of labelled DNA (≥20 bases) from unincorporated labelled nucleotides in a volume of 25 to 50 μl using spin column chromatography.
  • Columns are prepacked with Sephadex™ G-50 Grade pre-equilibrated in TE buffer with 0,05% Kathon CG/ICP Biocide
  • Product recovery will be at least 10% lower than that from ProbeQuant™ G-50 Micro Columns
  • Can also be used for buffer exchange/desalting of enzymatic reactions (10 to 100 μl) following heat denaturation and phenol extraction
Note: Not for use with ultrasalt-sensitive automated DNA sequencers.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty