Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Immobiline™ DryPlate for IEF
Supplier: Cytiva
Immobiline™
80-1128-28 80-1128-28
95017-717PK 1780.97 USD InStock
95017-717 CA95017-717L
Immobiline™ DryPlate for IEF
Electrophoresis Gels Precast Gels
Obtain reproducible IEF separations with dry, precast gels containing immobilized pH gradients.
  • Choose from narrow (1 pH unit) and medium (3 pH units) pH ranges
  • Handle stable, plastic-backed gels with confidence
Immobiline™ DryPlate precast gels contain preformed, immobilized, non-drifting pH gradients. The gels are rehydrated to a thickness of 0.5 mm in the reswelling cassette accessory before use.
Immobiline™ technology produces a stable pH gradient, allowing high voltages and resolution over narrow pH ranges, even for low mobility proteins that require long focusing times.
Ordering information: Immobiline™ sample application strips include 52 samples, 5–20µL and instructions.
Delivery information: Kit includes 50× filter paper, 3× experimental result form instructions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty