Would you like to log in again?
Protease Inhibitor Mix, Cytiva
Supplier: Cytiva
Ettan™
80-6501-23 80-6501-23
95017-782EA 454.3 USD InStock
95017-782 CA95017-782L
Protease Inhibitor Mix, Cytiva
Cell Culture Supplements and Additives
Sample preparation often requires the inhibition of protease activity. Protease inhibitor mix combines competitive and non competitive protease inhibitors, which protect proteins from proteolysis during purification from animal tissues, plant tissues, yeast and bacteria.
  • Protease inhibitor cocktail specifically developed for sample preparation in 2-D studies
  • No EDTA is used, which allows optimal nuclease activity for removing nucleic acids from samples
  • Contains a mixture of both competitive and noncompetitive protease inhibitors that inhibit serine, cysteine, and calpain proteases - optionally, EDTA may be added to inhibit metalloproteases
  • Optimised concentration for excellent inhibition of protease activities
  • Effectively inhibits over 95% of protease activity
Protease inhibitor mix combines competitive and non competitive protease inhibitors, which protect proteins from proteolysis during purification from animal tissues, plant tissues, yeast and bacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty