Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BrdU Mouse Monoclonal Antibody

Supplier: Cytiva
RPN202 RPN202
95019-028EA 1266.39 USD InStock
CA95019-028L 95019-028
Anti-BrdU Mouse Monoclonal Antibody
Antibodies
Anti-Bromodeoxyuridine (Anti-BRdU) antibody is supplied in the same formulation as the monoclonal antibody to BrdU in the Cell Proliferation Kit. The antibody/nuclease mixture is compatible with detection systems that incorporate fluorescently labelled antibodies and biotin-streptavidin detection reagents.

  • No need for chemical denaturation
  • Detection while maintaining cell morphology
  • Double immunocytochemical staining studies can be perfromed

Used in conjunction with RPN20, the reagent allows extended use of the kit where additional amounts of labeling reagent are required, such as in vivo studies.

The antibody contains a nuclease to produce single stranded DNA without the need for chemical denaturation. Cellular morphology is preserved allowing multiple labeling experiments to be carried out. The high-contrast stain produced has sensitivity comparable to autoradiographic methods.

Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Chemical

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty