Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Sheep Antibody (HRP (Horseradish Peroxidase))

Supplier: Cytiva

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

RPN4201 RPN4201
95019-218EA 1668.21 USD InStock
95019-218 CA95019-218L
Anti-IgG Sheep Antibody (HRP (Horseradish Peroxidase))
Antibodies
For use in ELISA systems
For ELISA, it is recommended that the conjugate should be diluted in the range 1:4000 to 1:8000
Amdex conjugates are already supplied as components of the Biotrak high-sensitivity cytokine ELISAs, complete with fully-optimized protocols
Easily soluble in cold and hot water

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty