Would you like to log in again?
Nutrient Broth, Himedia Laboratories
Supplier: HiMedia Laboratories
M002-500G M002-2.5KG M002-500G M002-5KG M002-5KG M002-2.5KG
89135-054EA 153.18 USD InStock
CA89135-054 CA95019-998 89135-054 CA95026-760 95026-760 95019-998
Nutrient Broth, Himedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Nutrient broth is used for the general cultivation of less fastidious microorganisms , can be enriched with blood or other biological fluids.
Certifications: Complies as per ISO 17025:2005.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty