Would you like to log in again?
MacConkey Agar Medium, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M008E-500G M008E-500G
95020-020EA 175.32 USD InStock
95020-020 CA95020-020
MacConkey Agar Medium, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and differentiation of lactose fermenting and lactose nonfermenting enteric bacteria and also for isolation of fecal streptococci.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty