Would you like to log in again?
Nutrient Agar with 1% Peptone, HiMedia
Supplier: HiMedia Laboratories
M012-2.5KG M012-500G M012-500G M012-2.5KG M012-5KG M012-5KG
89222-192EA 1112.81 USD InStock
89222-192 CA89222-190 89222-190 CA95020-036 95020-036 CA89222-192
Nutrient Agar with 1% Peptone, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Nutrient agar medium is used as a general purpose culture media.
  • Cream to yellow homogeneous free flowing powder
  • pH of 7.20 - 7.60
  • Firm, comparable with 1.5 % Agar gel
Nutrient agar medium is used as a general purpose culture media which may be used as enriched medium by incorporating 10 % blood or other biological fluid.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty