Would you like to log in again?
Deoxycholate Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M030-500G M030-500G
95020-090EA 182.34 USD InStock
CA95020-090 95020-090
Deoxycholate Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the direct differential count of coliforms in dairy products and for the isolation of enteric pathogens from rectal swabs, feces, and other pathological specimens.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty