Would you like to log in again?
Phenol Red Agar Base Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M053-500G M053-500G
95020-140EA 184.99 USD InStock
95020-140 CA95020-140
Phenol Red Agar Base Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
A basal medium to which carbohydrates may be added for use in fermentation studies of microorganisms.
  • Microbiological culture media
  • 500 g poly bottle
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty