Would you like to log in again?
Nutrient Gelatin Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M060S-500G M060S-2.5KG M060S-5KG M060S-5KG M060-2.5KG M060-5KG M060S-500G M060-2.5KG M060-500G M060-5KG M060-500G M060S-2.5KG
89222-368EA 1175.2 USD InStock
CA89222-368 CA89222-366 89222-368 95020-156 CA95020-154 95020-154 89222-362 CA95020-156 CA89222-364 89222-366 CA89222-362 89222-364
Nutrient Gelatin Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the detection of gelatin liquefaction by proteolytic microorganisms.
  • Microbiological culture media
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty