Would you like to log in again?
MacConkey Agar with CV, NaCl, and 0.15% Bile Salts, Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M081A-500G M081A-500G
95020-206EA 133.2 USD InStock
CA95020-206 95020-206
MacConkey Agar with CV, NaCl, and 0.15% Bile Salts, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Used to identify Enterobacteriaceae in the presence of coliforms and lactose nonfermenters from water, sewage, and food products.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty