Would you like to log in again?
MacConkey Agar without CV and NaCl, with 0.5% Bile Salts, Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M082A-500G M082A-500G
95020-212EA 193.45 USD InStock
CA95020-212 95020-212
MacConkey Agar without CV and NaCl, with 0.5% Bile Salts, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and differentiation of enteric bacteria, restricting swarming of Proteus species from specimens such as urine, which may contain large numbers of Proteus species as well as potentially pathogenic gram-positive organisms.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty