Would you like to log in again?
Eijkman Lactose Broth Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M086-500G M086-500G
95020-226EA 219.1 USD InStock
95020-226 CA95020-226
Eijkman Lactose Broth Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the detection and differentiation of Escherichia coli from other coliforms based on the ability to grow and liberate gas from lactose.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty