Would you like to log in again?
Dichloran Medium Base with Rose Bengal, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1000-500G M1000-500G
95020-258EA 219.82 USD InStock
95020-258 CA95020-258
Dichloran Medium Base with Rose Bengal, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and enumeration of yeasts and molds associated with food spoilage.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty