Would you like to log in again?
Sporulation Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1018-5KG M1018-2.5KG M1018-500G M1018-500G M1018-2.5KG M1018-5KG
89222-610EA 1291.1 USD InStock
CA89222-610 89222-610 95020-298 89222-608 CA95020-298 CA89222-608
Sporulation Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Sporulation broth is used for promoting sporulation in Bacillus subtilis.
  • Cream to light yellow colored
  • Homogeneous, free flowing powder
  • Reaction of 1.58% w/v aqueous solution at 25 °C
Caution: Store below 30 °C in tightly closed container and the prepared medium at 2 to 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty