Would you like to log in again?
M17 Agar without Lactose, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1019-500G M1019-500G
95020-300EA 476.37 USD InStock
CA95020-300 95020-300
M17 Agar without Lactose, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
On addition of lactose, this medium can be used for cultivation and isolation of lactic streptococci.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty