Would you like to log in again?
Bile Esculin Agar with Kanamycin, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1035-500G M1035-500G
95020-338EA 274.63 USD InStock
CA95020-338 95020-338
Bile Esculin Agar with Kanamycin, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and presumptive identification of bacteria of the Bacteroides fragilis group from mixed flora.
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty