Would you like to log in again?
Cooked Meat Medium with Glucose, Hemin, and Vitamin K, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1040-500G M1040-500G
95020-348EA 444.01 USD InStock
CA95020-348 95020-348
Cooked Meat Medium with Glucose, Hemin, and Vitamin K, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of aerobes and anaerobes, especially pathogenic Clostridia.
  • Microbiological culture media
  • 500 g poly bottles
Also for the maintenance of stock cultures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty