Would you like to log in again?
Esculin Iron Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1044-500G M1044-500G
95020-356EA 284.31 USD InStock
CA95020-356 95020-356
Esculin Iron Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For verifying enterococcal colonies on membrane filters through which water samples have been filtered and which have been incubated on m-Enterococcus agar, modified.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty