Would you like to log in again?
Coliform PA Broth Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1051-500G M1051-500G
95020-374EA 211.08 USD InStock
CA95020-374 95020-374
Coliform PA Broth Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the determination of the presence or absence of coliform bacteria in examination of pollution in treated water from water treatment plants or distribution systems.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty