Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brain Heart Infusion Agar with 3.0% Agar, Dehydrated Culture Media, HiMedia Laboratories

Supplier: HiMedia Laboratories
M1069-500G M1069-500G
95020-414EA 265.53 USD InStock
CA95020-414 95020-414
Brain Heart Infusion Agar with 3.0% Agar, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of microorganisms when 3.0% agar gel is required.

  • Microbiological culture media
  • Packaged in 500 g poly bottles

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty