Would you like to log in again?
Starch Agar, HiMedia Laboratories
Supplier: HiMedia Laboratories
M107-500G M107-5KG M107-500G M107-5KG M107-2.5KG M107-2.5KG
89222-824EA 651.26 USD InStock
89222-824 CA89222-838 89222-838 CA89222-824 CA95020-426 95020-426
Starch Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Starch agar is used for the detection of starch hydrolyzing microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty