Would you like to log in again?
Glutamate Starch-Phenol Red Agar Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1089-500G M1089-500G
95020-460EA 216.81 USD InStock
95020-460 CA95020-460
Glutamate Starch-Phenol Red Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the detection of Pseudomonas and Aeromonas species in foodstuffs, waste water, and equipment in the food industry
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty