Would you like to log in again?
M-Enterococcus Agar Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1108-500G M1108-500G
95020-490EA 183.69 USD InStock
CA95020-490 95020-490
M-Enterococcus Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and enumeration of enterococci in water, sewage, food and other materials by membrane filter technique as well as direct plating of specimens.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty