Would you like to log in again?
Urea Agar Base (Christensen), HiMedia Laboratories
Supplier: HiMedia Laboratories
M112-500G M112-500G
95020-516EA 192.45 USD InStock
CA95020-516 95020-516
Urea Agar Base (Christensen), HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Urea agar base with the addition of urea is used for the detection of urease production, particularly by members of the genus Proteus.
  • Autoclavable
  • Store below 30 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty