Would you like to log in again?
Columbia Blood Agar Base with Hemin, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1133-500G M1133-500G
95020-548EA 523.96 USD InStock
CA95020-548 95020-548
Columbia Blood Agar Base with Hemin, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
An efficient and enriched base for preparation of blood agar or chocolate agar, and for various selective and identification media.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty