Would you like to log in again?
Oxytetra Glucose Yeast Agar Base with Biotin, Dehydrated, HiMedia Laboratories
Supplier: HiMedia
M1136-500G M1136-500G
95020-552EA 244.61 USD InStock
95020-552 CA95020-552
Oxytetra Glucose Yeast Agar Base with Biotin, Dehydrated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and enumeration of yeast and molds from foodstuffs.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty