Would you like to log in again?
WL Nutrient Agar, HiMedia
Supplier: HiMedia Laboratories
M115-2.5KG M115-5KG M115-500G M115-500G
89223-136EA 1012.3 USD InStock
89223-136 95020-592 CA95020-592 89223-134
WL Nutrient Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
WL Nutrient Agar is recommended for selective isolation and enumeration of bacteria encountered in breweries and industrial fermentations.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty